MAT379362193 Matrix Scientific

Ethyl 2-methylbenzoylformate

  • MDL number MFCD01311167
  • Cas Number : 66644-67-5
  • Molecular Weight : 192.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/086348.html

Safety Information

MSDS http://www.matrixscientific.com/086348.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart