MAT379362271 Matrix Scientific

cis-1,3-Cyclopentanedicarboxylic anhydride

  • MDL number MFCD00077242
  • Cas Number : 6054-16-6
  • Molecular Weight : 140.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 158-160
Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/086426.html

Safety Information

MSDS http://www.matrixscientific.com/086426.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart