MAT379362288 Matrix Scientific

trans-2-(2-Thiomethylbenzoyl)cyclopentane-1-carboxylic acid

  • MDL number MFCD01311238
  • Cas Number : 1134700-56-3
  • Molecular Weight : 264.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 135-137
Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/086443.html

Safety Information

MSDS http://www.matrixscientific.com/086443.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart