MAT379362298 Matrix Scientific

cis-2-(2-Methoxybenzoyl)cyclohexane-1-carboxylic acid

  • MDL number MFCD01311249
  • Cas Number : 733742-61-5
  • Molecular Weight : 262.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 141-149
Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/086453.html

Safety Information

MSDS http://www.matrixscientific.com/086453.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart