MAT379362300 Matrix Scientific

trans-2-Benzoylcyclohexane-1-carboxylic acid

  • MDL number MFCD00800520
  • Cas Number : 3586-84-3
  • Molecular Weight : 232.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 149-151
Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/086455.html

Safety Information

MSDS http://www.matrixscientific.com/086455.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart