MAT379362303 Matrix Scientific

cis-3-Benzoylcyclohexane-1-carboxylic acid

  • MDL number MFCD01311278
  • Cas Number : 733742-86-4
  • Molecular Weight : 232.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 105-110
Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/086458.html

Safety Information

MSDS http://www.matrixscientific.com/086458.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart