MAT379362309 Matrix Scientific

trans-4-(4-Cyanobenzoyl)cyclohexane-1-carboxylic acid

  • MDL number MFCD01319775
  • Cas Number : 736136-09-7
  • Molecular Weight : 257.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 197-200
Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/086464.html

Safety Information

MSDS http://www.matrixscientific.com/086464.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart