MAT379362340 Matrix Scientific

4-Chloro-2'-methoxybenzophenone

  • MDL number MFCD01311535
  • Cas Number : 78589-10-3
  • Molecular Weight : 246.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 83-85
Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/086517.html

Safety Information

MSDS http://www.matrixscientific.com/086517.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart