MAT379362342 Matrix Scientific

3-Methoxybenzophenone

  • MDL number MFCD01319936
  • Cas Number : 6136-67-0
  • Molecular Weight : 212.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 36-38
Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/086519.html

Safety Information

MSDS http://www.matrixscientific.com/086519.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart