MAT379362347 Matrix Scientific

4-Cyano-4'-methoxybenzophenone

  • MDL number MFCD01311594
  • Cas Number : 27645-60-9
  • Molecular Weight : 237.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 131-133
Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/086524.html

Safety Information

MSDS http://www.matrixscientific.com/086524.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart