MAT379362351 Matrix Scientific

4-Bromo-4'-methoxybenzophenone

  • MDL number MFCD00228667
  • Cas Number : 54118-75-1
  • Molecular Weight : 291.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 156-159
Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/086528.html

Safety Information

MSDS http://www.matrixscientific.com/086528.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart