MAT379362367 Matrix Scientific

2-Ethoxycarbonyl-4'-nitrobenzophenone

  • MDL number MFCD02260407
  • Cas Number : 760192-93-6
  • Molecular Weight : 299.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 106-108
Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/086544.html

Safety Information

MSDS http://www.matrixscientific.com/086544.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart