MAT379362380 Matrix Scientific

4-Acetoxy-4'-nitrobenzophenone

  • MDL number MFCD07698961
  • Cas Number : 890099-64-6
  • Molecular Weight : 285.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 130-131
Purity 90
Country of Origin USA
Product Link http://www.matrixscientific.com/086557.html

Safety Information

MSDS http://www.matrixscientific.com/086557.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart