MAT379362381 Matrix Scientific

4-Acetoxy-3',4'-dichlorobenzophenone

  • MDL number MFCD07698987
  • Cas Number : 890100-13-7
  • Molecular Weight : 309.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 105-107
Purity 90
Country of Origin USA
Product Link http://www.matrixscientific.com/086558.html

Safety Information

MSDS http://www.matrixscientific.com/086558.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart