MAT379362383 Matrix Scientific

3,3',5,5'-Tetrachlorobenzophenone

  • MDL number MFCD01763607
  • Cas Number : 92758-86-6
  • Molecular Weight : 320

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 161-164
Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/086560.html

Safety Information

MSDS http://www.matrixscientific.com/086560.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart