MAT379362384 Matrix Scientific

3,3',5,5'-Tetrafluorobenzophenone

  • MDL number MFCD06201523
  • Cas Number : 139911-09-4
  • Molecular Weight : 254.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 85-87
Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/086561.html

Safety Information

MSDS http://www.matrixscientific.com/086561.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart