MAT379362387 Matrix Scientific

4-Bromo-2'-methylbenzophenone

  • MDL number MFCD09801661
  • Cas Number : 27428-59-7
  • Molecular Weight : 275.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 40-41
Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/086564.html

Safety Information

MSDS http://www.matrixscientific.com/086564.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart