MAT379362828 Matrix Scientific

6-Oxodecanoic acid

  • MDL number MFCD01320163
  • Cas Number : 4144-60-9
  • Molecular Weight : 186.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 44-46
Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/087007.html

Safety Information

MSDS http://www.matrixscientific.com/087007.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart