MAT379362971 Matrix Scientific

2'-Chloro-3-(1,3-dioxan-2-yl)propiophenone

  • MDL number MFCD02261794
  • Cas Number : 898785-82-5
  • Molecular Weight : 254.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/087149.html

Safety Information

MSDS http://www.matrixscientific.com/087149.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart