MAT379362974 Matrix Scientific

4-(5,5-Dimethyl-1,3-dioxan-2-yl)-2'-fluorobutyrophenone

  • MDL number MFCD03844241
  • Cas Number : 898786-04-4
  • Molecular Weight : 280.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 60-61
Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/087152.html

Safety Information

MSDS http://www.matrixscientific.com/087152.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart