MAT379362975 Matrix Scientific

3-(1,3-Dioxan-2-yl)-4'-trifluoromethylpropiophenone

  • MDL number MFCD02261808
  • Cas Number : 898786-59-9
  • Molecular Weight : 288.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 46-47
Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/087153.html

Safety Information

MSDS http://www.matrixscientific.com/087153.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart