MAT379362976 Matrix Scientific

2-(1,3-Dioxolan-2-yl)-1-(3-fluoro-phenyl)-ethanone

  • MDL number MFCD18433440
  • Cas Number : 1166996-40-2
  • Molecular Weight : 210.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/087154.html

Safety Information

MSDS http://www.matrixscientific.com/087154.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart