MAT379362977 Matrix Scientific

(1,3-Dioxolan-2-yl)-acetic acid methyl ester

  • MDL number MFCD18433502
  • Cas Number : 107599-68-8
  • Molecular Weight : 146.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Boiling point 94-96�Torr
Product Link http://www.matrixscientific.com/087155.html

Safety Information

MSDS http://www.matrixscientific.com/087155.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart