MAT379362997 Matrix Scientific

5-Bromo-1-fluoro-3-methyl-2-nitro-benzene

  • MDL number MFCD22566379
  • Cas Number : 1224629-03-1
  • Molecular Weight : 234.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 55-60
Purity 95
Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/087180.html

Safety Information

MSDS http://www.matrixscientific.com/087180.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart