MAT386586669 Matrix Scientific

5-Bromo-1-chloro-3-fluoro-2-(trifluoromethoxy)benzene

  • MDL number MFCD22580729
  • Cas Number : 1417566-38-1
  • Molecular Weight : 293.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Product Link https://www.matrixscientific.com/087318.html

Safety Information

hcodes Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart