MAT386586701 Matrix Scientific

2-Fluoro-4-(trifluoromethoxy)benzonitrile

  • MDL number MFCD16652437
  • Cas Number : 1240256-78-3
  • Molecular Weight : 205.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/087350.html

Safety Information

MSDS http://www.matrixscientific.com/087350.html
pictograms Toxic
Availability details for
Quantity
  • Availability
  • Add to Cart