MAT386586754 Matrix Scientific

1-Bromo-4-[chloro(difluoro)methoxy]benzene

  • MDL number MFCD22580792
  • Cas Number : 112556-13-5
  • Molecular Weight : 257.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/087403.html

Safety Information

MSDS http://www.matrixscientific.com/087403.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart