MAT386586848 Matrix Scientific

2-Bromo-1,3-difluoro-5-(trifluoromethoxy)benzene

  • MDL number MFCD22580861
  • Cas Number : 1417568-00-3
  • Molecular Weight : 276.99

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/087497.html

Safety Information

MSDS http://www.matrixscientific.com/087497.html
pictograms Irritant/Refrigerate
Availability details for
Quantity
  • Availability
  • Add to Cart