MAT388128024 Matrix Scientific

2-Chlorothiazolo[4,5-b]pyridine hydrochloride

  • MDL number MFCD20482877
  • Cas Number : 152170-30-4
  • Molecular Weight : 207.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/073760.html

Safety Information

MSDS http://www.matrixscientific.com/073760.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart