MAT388128104 Matrix Scientific

Dimethyl (morpholin-4-yl)phosphonate

  • MDL number MFCD00514647
  • Cas Number : 597-25-1
  • Molecular Weight : 195.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/087895.html

Safety Information

MSDS http://www.matrixscientific.com/087895.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart