MAT388366984 Matrix Scientific

[1-Methyl-5-(trifluoromethyl)-1H-benzimidazol-2-yl]methylamine hydrochloride

  • MDL number MFCD18904335
  • Cas Number : 1281872-57-8
  • Molecular Weight : 265.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/035855.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/035855.html
Availability details for
Quantity
  • Availability
  • Add to Cart