MAT388367372 Matrix Scientific

1-Chloro-2-[(4-chlorobenzyl)oxy]-4-fluoro-5-nitrobenzene

  • MDL number MFCD18384917
  • Cas Number : 1355170-75-0
  • Molecular Weight : 316.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/088354.html

Safety Information

MSDS http://www.matrixscientific.com/088354.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart