MAT388367483 Matrix Scientific

2-(4-Formylphenoxy)nicotinonitrile

  • MDL number MFCD08741856
  • Cas Number : 919731-40-1
  • Molecular Weight : 224.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/088465.html

Safety Information

MSDS http://www.matrixscientific.com/088465.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart