MAT388367664 Matrix Scientific

4-Bromo-1-methyl-1H-pyrazole-3-carbonitrile

  • MDL number MFCD00103545
  • Cas Number : 287922-71-8
  • Molecular Weight : 186.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/088646.html

Safety Information

MSDS http://www.matrixscientific.com/088646.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart