MAT388367995 Matrix Scientific

N-(tert-Butyl)-1,2,3,4-tetrahydroisoquinoline-3-carboxamide

  • MDL number MFCD01315995
  • Cas Number : 149182-72-9
  • Molecular Weight : 232.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/088975.html

Safety Information

MSDS http://www.matrixscientific.com/088975.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart