MAT388368130 Matrix Scientific

(3,5-Dichlorophenyl)methanesulfonyl chloride

  • MDL number MFCD01631928
  • Cas Number : 163295-70-3
  • Molecular Weight : 259.54

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 105-110
Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/089110.html

Safety Information

MSDS http://www.matrixscientific.com/089110.html
pictograms Corrosive/Water Reactive
Availability details for
Quantity
  • Availability
  • Add to Cart