MAT390161273 Matrix Scientific

4-Chloro-2-(difluoromethyl)pyridine hydrochloride

  • MDL number MFCD23701768
  • Cas Number : 1553437-88-9
  • Molecular Weight : 200.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/071594.html

Safety Information

MSDS http://www.matrixscientific.com/071594.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart