MAT390161277 Matrix Scientific

5-Methylpyridine-2-carboxylic acid hydrochloride

  • MDL number MFCD20040409
  • Cas Number : 177359-60-3
  • Molecular Weight : 173.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point >300
Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/071369.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/071369.html
Availability details for
Quantity
  • Availability
  • Add to Cart