MAT390161278 Matrix Scientific

5-Methyl-4-(pyrrolidin-1-ylmethyl)isoxazole-3-carboxylic acid hydrochloride

  • MDL number MFCD11502958
  • Cas Number : 893750-02-2
  • Molecular Weight : 246.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/036519.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/036519.html
Availability details for
Quantity
  • Availability
  • Add to Cart