MAT390500711 Matrix Scientific

2,3-Dihydro-1H-inden-1-ylamine hydrochloride

  • MDL number MFCD00054294
  • Cas Number : 34698-41-4
  • Molecular Weight : 169.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/037567.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/037567.html
Availability details for
Quantity
  • Availability
  • Add to Cart