MAT390501223 Matrix Scientific

Atazanavir sulfate

  • MDL number MFCD08067748
  • Cas Number : 229975-97-7
  • Molecular Weight : 802.95

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/089877.html

Safety Information

MSDS http://www.matrixscientific.com/089877.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart