MAT390501315 Matrix Scientific

6-Bromo-2-naphthaldehyde

  • MDL number MFCD09842469
  • Cas Number : 170737-46-9
  • Molecular Weight : 235.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/089969.html

Safety Information

MSDS http://www.matrixscientific.com/089969.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart