MAT390501703 Matrix Scientific

2-Amino-1,5-naphthalenedisulfonic acid

  • MDL number MFCD00021512
  • Cas Number : 117-62-4
  • Molecular Weight : 303.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/090360.html

Safety Information

MSDS http://www.matrixscientific.com/090360.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart