MAT390501876 Matrix Scientific

(1S,2R,3S,5R)-3-(Benzyloxy)-2-((benzyloxy)-methyl)-6-oxabicyclo[3.1.0]hexane

  • MDL number MFCD09750952
  • Cas Number : 110567-22-1
  • Molecular Weight : 310.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/090533.html

Safety Information

MSDS http://www.matrixscientific.com/090533.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart