MAT390502035 Matrix Scientific

3-(3-Iodophenyl)acrylic acid

  • MDL number MFCD00464508
  • Cas Number : 41070-12-6
  • Molecular Weight : 274.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/090694.html

Safety Information

MSDS http://www.matrixscientific.com/090694.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart