MAT390502296 Matrix Scientific

N1-(2-(4-(2-Aminoethyl)piperazin-1-yl)ethyl)ethane-1,2-diamine

  • MDL number MFCD01941014
  • Cas Number : 31295-54-2
  • Molecular Weight : 215.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Ambient/argon
Country of Origin USA
Product Link http://www.matrixscientific.com/090955.html

Safety Information

MSDS http://www.matrixscientific.com/090955.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart