MAT390502522 Matrix Scientific

4,4'-(1,4-Phenylenebis(ethene-2,1-diyl))dibenzonitrile

  • MDL number MFCD00369832
  • Cas Number : 13001-40-6
  • Molecular Weight : 332.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/091188.html

Safety Information

MSDS http://www.matrixscientific.com/091188.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart