MAT390502576 Matrix Scientific

(4-Bromonaphthalen-1-yl)methanamine hydrochloride

  • MDL number MFCD08706223
  • Cas Number : 578029-09-1
  • Molecular Weight : 272.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/091242.html

Safety Information

MSDS http://www.matrixscientific.com/091242.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart