MAT390502912 Matrix Scientific

N-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-2-(trifluoromethyl)phenyl)acetamide

  • MDL number MFCD11855983
  • Cas Number : 1150271-66-1
  • Molecular Weight : 329.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/091578.html

Safety Information

MSDS http://www.matrixscientific.com/091578.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart