MAT390503313 Matrix Scientific

2-(4-Fluoro-3-methylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

  • MDL number MFCD18729904
  • Cas Number : 1001200-60-7
  • Molecular Weight : 236.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/091981.html

Safety Information

MSDS http://www.matrixscientific.com/091981.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart